siRNA Details
Gene Symbol: Zfp512b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107809.1 | NP_001101279.1 | 5 | 917 | ||
XM_008762511.1 | XP_008760733.1 | 5 | 958 | |||
XM_008762512.1 | XP_008760734.1 | 4 | 811 | |||
XM_008762513.1 | XP_008760735.1 | 4 | 589 |
