siRNA Details
| Gene Symbol: Zfp512b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107809.1 | NP_001101279.1 | 6 | 1398 | ||
| XM_008762511.1 | XP_008760733.1 | 6 | 1439 | |||
| XM_008762512.1 | XP_008760734.1 | 5 | 1292 | |||
| XM_008762513.1 | XP_008760735.1 | 5 | 1070 | |||

Zfp512b
Genomic Map