siRNA Details
| Gene Symbol: Pcmtd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107810.1 | NP_001101280.2 | 7 | 1142 | ||
| GenBank mRNA | BC168892.1 | - | 1 | 397 | 7444809 | |
| FQ211587.1 | - | 7 | 1141 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcmtd2| Gene Symbol: Pcmtd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107810.1 | NP_001101280.2 | 7 | 1142 | ||
| GenBank mRNA | BC168892.1 | - | 1 | 397 | 7444809 | |
| FQ211587.1 | - | 7 | 1141 | |||