siRNA Details
| Gene Symbol: Pcmtd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107810.1 | NP_001101280.2 | 5 | 765 | ||
| GenBank mRNA | FQ211587.1 | - | 5 | 764 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcmtd2| Gene Symbol: Pcmtd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107810.1 | NP_001101280.2 | 5 | 765 | ||
| GenBank mRNA | FQ211587.1 | - | 5 | 764 | ||