siRNA Details
Gene Symbol: Metap1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107812.2 | NP_001101282.2 | 6, 7 | 777 | ||
XM_008761908.1 | XP_008760130.1 | 5, 6 | 622 | |||
GenBank mRNA | BC167092.1 | - | 6, 7 | 704 | ||
DY575699.1 | - | 5, 6 | 654 |
