siRNA Details
| Gene Symbol: Nelfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107817.2 | NP_001101287.2 | 8 | 1270 | ||
| GenBank mRNA | BC160853.1 | - | 7 | 1392 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nelfb| Gene Symbol: Nelfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107817.2 | NP_001101287.2 | 8 | 1270 | ||
| GenBank mRNA | BC160853.1 | - | 7 | 1392 | ||