siRNA Details
| Gene Symbol: Ndor1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107818.1 | NP_001101288.1 | 5 | 519 | ||
| XM_006233612.2 | XP_006233674.1 | 4 | 811 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ndor1| Gene Symbol: Ndor1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107818.1 | NP_001101288.1 | 5 | 519 | ||
| XM_006233612.2 | XP_006233674.1 | 4 | 811 | |||