siRNA Details
| Gene Symbol: Ak8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004266.1 | NP_001004266.1 | 4, 5 | 356 | ||
| XM_006233794.2 | XP_006233856.1 | 5, 6 | 392 | |||
| GenBank mRNA | BC079446.1 | - | 4, 5 | 356 | 7115585 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ak8| Gene Symbol: Ak8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004266.1 | NP_001004266.1 | 4, 5 | 356 | ||
| XM_006233794.2 | XP_006233856.1 | 5, 6 | 392 | |||
| GenBank mRNA | BC079446.1 | - | 4, 5 | 356 | 7115585 | |