siRNA Details
Gene Symbol: Zer1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100707.1 | NP_001094177.1 | 11 | 2116 | ||
XM_008761652.1 | XP_008759874.1 | 11 | 2027 | |||
XM_008761653.1 | XP_008759875.1 | 11 | 2021 | |||
XM_008761654.1 | XP_008759876.1 | 11 | 2027 | |||
XM_008761655.1 | XP_008759877.1 | 11 | 1878 | |||
XM_008761656.1 | XP_008759878.1 | 11 | 2264 | |||
XM_008761657.1 | XP_008759879.1 | 11 | 2213 | |||
GenBank mRNA | FQ233933.1 | - | 1 | 69 |
