siRNA Details
Gene Symbol: Zer1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100707.1 | NP_001094177.1 | 4 | 1115 | ||
XM_008761652.1 | XP_008759874.1 | 4 | 1065 | |||
XM_008761653.1 | XP_008759875.1 | 4 | 1065 | |||
XM_008761654.1 | XP_008759876.1 | 4 | 1065 | |||
XM_008761655.1 | XP_008759877.1 | 4 | 916 | |||
XM_008761656.1 | XP_008759878.1 | 4 | 1302 | |||
XM_008761657.1 | XP_008759879.1 | 4 | 1251 |
