siRNA Details
| Gene Symbol: Lrrc8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024782.1 | NP_001019953.1 | 3 | 454 | ||
| XM_006233814.2 | XP_006233876.1 | 1 | 300 | |||
| XM_008761660.1 | XP_008759882.1 | 1 | 300 | |||
| GenBank mRNA | BC097371.1 | - | 3 | 454 | 7444090 | |

Lrrc8a
Genomic Map