siRNA Details
| Gene Symbol: Fibcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107829.1 | NP_001101299.1 | 2 | 538 | ||
| Gene Symbol: NEWGENE_1309097 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761689.1 | XP_008759911.1 | 2 | 514 | ||

Fibcd1
Genomic Map