siRNA Details
| Gene Symbol: Lhx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107837.1 | NP_001101307.1 | 6, 7 | 825 | ||
| XM_006234066.2 | XP_006234128.1 | 6, 7 | 1026 | |||
| XM_008761753.1 | XP_008759975.1 | 6, 7 | 1087 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lhx6| Gene Symbol: Lhx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107837.1 | NP_001101307.1 | 6, 7 | 825 | ||
| XM_006234066.2 | XP_006234128.1 | 6, 7 | 1026 | |||
| XM_008761753.1 | XP_008759975.1 | 6, 7 | 1087 | |||