siRNA Details
Gene Symbol: Rc3h2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107839.1 | NP_001101309.1 | 8 | 1253 | ||
XM_008761754.1 | XP_008759976.1 | 8 | 1354 | |||
XM_008761755.1 | XP_008759977.1 | 8 | 1354 |
