siRNA Details
| Gene Symbol: Rabgap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107841.1 | NP_001101311.1 | 5, 6 | 991 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rabgap1| Gene Symbol: Rabgap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107841.1 | NP_001101311.1 | 5, 6 | 991 | ||