siRNA Details
| Gene Symbol: Galnt11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271319.1 | NP_001258248.1 | 5 | 754 | ||
| NM_199393.2 | NP_955425.2 | 4 | 671 | |||
| XM_006235922.2 | XP_006235984.1 | 3 | 812 | |||
| XM_006235923.2 | XP_006235985.1 | 4 | 1164 | |||
| GenBank mRNA | BC062004.1 | - | 4 | 605 | 5599809 | |
| FQ223626.1 | - | 5 | 754 | |||

Galnt11
Genomic Map