siRNA Details
| Gene Symbol: Kmt2e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100851.1 | NP_001094321.1 | 16 | 3219 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kmt2e| Gene Symbol: Kmt2e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100851.1 | NP_001094321.1 | 16 | 3219 | ||