siRNA Details
| Gene Symbol: Pon3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004086.1 | NP_001004086.1 | 6 | 568 | ||
| GenBank mRNA | BC079466.1 | - | 6 | 568 | 7122531 | |
| FQ218322.1 | - | 3 | 307 | |||
| FQ218352.1 | - | 3 | 307 | |||
| FQ218880.1 | - | 3 | 307 | |||
| FQ218895.1 | - | 3 | 308 | |||
| FQ219524.1 | - | 3 | 307 | |||

Pon3
Genomic Map