siRNA Details
| Gene Symbol: Stard8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107849.2 | NP_001101319.2 | 4 | 1470 | ||
| XM_006257073.2 | XP_006257135.1 | 5 | 2978 | |||
| XM_006257074.2 | XP_006257136.1 | 6 | 1710 | |||
| XM_006257075.2 | XP_006257137.1 | 5 | 1594 | |||

Stard8
Genomic Map