siRNA Details
| Gene Symbol: Tmem106b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004267.1 | NP_001004267.1 | 7, 8 | 755 | ||
| GenBank mRNA | BC079127.1 | - | 7, 8 | 755 | 7127112 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem106b| Gene Symbol: Tmem106b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004267.1 | NP_001004267.1 | 7, 8 | 755 | ||
| GenBank mRNA | BC079127.1 | - | 7, 8 | 755 | 7127112 | |