siRNA Details
| Gene Symbol: RGD1306271 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006236338.2 | XP_006236400.1 | 2 | 2087 | ||
| XM_006236341.2 | XP_006236403.1 | 2 | 3730 | |||
| XM_008762846.1 | XP_008761068.1 | 2 | 3301 | |||
| XM_008762847.1 | XP_008761069.1 | 2 | 2047 | |||
| XM_231616.8 | XP_231616.5 | 2 | 2087 | |||

RGD1306271
Genomic Map