siRNA Details
Gene Symbol: Zfp956 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014056.1 | NP_001014078.1 | 4, 5 | 751 | ||
XM_006236417.2 | XP_006236479.1 | 5, 6 | 1079 | |||
XM_006236418.2 | XP_006236480.1 | 6, 7 | 1112 | |||
GenBank mRNA | BC089951.1 | - | 4, 5 | 751 | 7312211 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
