siRNA Details
| Gene Symbol: Zfp746 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100852.1 | NP_001094322.1 | 7 | 2176 | ||
| XM_006236419.2 | XP_006236481.1 | 7 | 2615 | |||
| XM_008762885.1 | XP_008761107.1 | 6 | 4507 | |||
| XM_008762886.1 | XP_008761108.1 | 6 | 4504 | |||
| XM_008762887.1 | XP_008761109.1 | 7 | 2610 | |||
| XM_008762888.1 | XP_008761110.1 | 5 | 4381 | |||
| GenBank mRNA | BC087150.1 | - | 1 | 1037 | ||

Zfp746
Genomic Map