siRNA Details
Gene Symbol: Nfe2l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_231763.8 | XP_231763.6 | 1938 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nfe2l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_231763.8 | XP_231763.6 | 1938 |