siRNA Details
| Gene Symbol: Avl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002729314.4 | XP_002729360.1 | 10 | 1488 | ||
| XM_006236564.2 | XP_006236626.1 | 10 | 1490 | |||
| XM_006236565.2 | XP_006236627.1 | 10 | 1490 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Avl9| Gene Symbol: Avl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002729314.4 | XP_002729360.1 | 10 | 1488 | ||
| XM_006236564.2 | XP_006236626.1 | 10 | 1490 | |||
| XM_006236565.2 | XP_006236627.1 | 10 | 1490 | |||