siRNA Details
| Gene Symbol: Kbtbd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107861.1 | NP_001101331.1 | 4 | 1296 | ||
| XM_006236547.2 | XP_006236609.1 | 4 | 1607 | |||
| XM_006236548.2 | XP_006236610.1 | 4 | 1451 | |||
| XM_006236549.2 | XP_006236611.1 | 4 | 1868 | |||
| XM_008762892.1 | XP_008761114.1 | 4 | 1973 | |||
| XM_008762893.1 | XP_008761115.1 | 5 | 1566 | |||
| XM_008762894.1 | XP_008761116.1 | 4 | 1341 | |||

Kbtbd2
Genomic Map