siRNA Details
| Gene Symbol: Smarcad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107864.2 | NP_001101334.2 | 12 | 1757 | ||
| XM_006236599.1 | XP_006236661.1 | 13 | 1840 | |||
| XM_006236600.2 | XP_006236662.1 | 13 | 1906 | |||
| XM_006236601.2 | XP_006236663.1 | 12 | 2996 | |||
| XM_008762979.1 | XP_008761201.1 | 12 | 3771 | |||

Smarcad1
Genomic Map