siRNA Details
| Gene Symbol: Smarcad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107864.2 | NP_001101334.2 | 22 | 2995 | ||
| XM_006236599.1 | XP_006236661.1 | 23 | 3078 | |||
| XM_006236600.2 | XP_006236662.1 | 23 | 3144 | |||
| XM_006236601.2 | XP_006236663.1 | 22 | 4234 | |||
| XM_008762979.1 | XP_008761201.1 | 22 | 5009 | |||

Smarcad1
Genomic Map