siRNA Details
| Gene Symbol: Cyp26b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181087.2 | NP_851601.1 | 6 | 1326 | ||
| GenBank mRNA | AY245532.2 | - | 6 | 1326 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp26b1| Gene Symbol: Cyp26b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181087.2 | NP_851601.1 | 6 | 1326 | ||
| GenBank mRNA | AY245532.2 | - | 6 | 1326 | ||