siRNA Details
Gene Symbol: Tia1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012096.1 | NP_001012096.1 | 11, 12 | 1184 | ||
XM_006236828.2 | XP_006236890.1 | 12, 13 | 1294 | |||
XM_006236829.2 | XP_006236891.1 | 12, 13 | 1293 | |||
XM_006236830.2 | XP_006236892.1 | 11, 12 | 1261 | |||
XM_006236831.2 | XP_006236893.1 | 12, 13 | 1091 | |||
GenBank mRNA | BC087064.1 | - | 11, 12 | 1184 | 7132038 |
