siRNA Details
| Gene Symbol: Gmcl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010956.2 | NP_001010956.1 | 7 | 990 | ||
| NM_001033931.1 | NP_001029103.1 | 7 | 990 | |||
| XM_006236832.2 | XP_006236894.1 | 6 | 939 | |||
| GenBank mRNA | AY862307.1 | - | 7 | 817 | ||
| AY862308.1 | - | 7 | 785 | |||

Gmcl1
Genomic Map