siRNA Details
| Gene Symbol: Pdzrn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271251.1 | NP_001258180.1 | 10 | 1960 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdzrn3| Gene Symbol: Pdzrn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271251.1 | NP_001258180.1 | 10 | 1960 | ||