siRNA Details
| Gene Symbol: Adipor2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037979.1 | NP_001033068.1 | 7 | 1125 | ||
| XM_006237183.1 | XP_006237245.1 | 8 | 1431 | |||
| XM_006237184.2 | XP_006237246.1 | 8 | 1302 | |||
| XM_006237185.2 | XP_006237247.1 | 8 | 1317 | |||
| XM_006237186.2 | XP_006237248.1 | 8 | 1446 | |||
| GenBank mRNA | AY724514.1 | - | 8 | 1385 | ||
| DQ148392.1 | - | 7 | 1125 | |||
| FQ232508.1 | - | 8 | 1404 | |||

Adipor2
Genomic Map