siRNA Details
| Gene Symbol: Clec2e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177689.1 | NP_001171160.1 | 2 | 117 | ||
| XM_008763336.1 | XP_008761558.1 | 3 | 2209 | |||
| GenBank mRNA | FJ404473.1 | - | 2 | 153 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clec2e| Gene Symbol: Clec2e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177689.1 | NP_001171160.1 | 2 | 117 | ||
| XM_008763336.1 | XP_008761558.1 | 3 | 2209 | |||
| GenBank mRNA | FJ404473.1 | - | 2 | 153 | ||