siRNA Details
| Gene Symbol: Gpr19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080579.1 | NP_542146.1 | 2 | 771 | ||
| XM_006237520.2 | XP_006237582.1 | 5 | 2954 | |||
| XM_006237521.2 | XP_006237583.1 | 5 | 1235 | |||
| XM_006237522.2 | XP_006237584.1 | 3 | 780 | |||
| XM_006237523.2 | XP_006237585.1 | 3 | 1027 | |||
| XM_006237524.1 | XP_006237586.1 | 3 | 937 | |||
| XM_008763379.1 | XP_008761601.1 | 4 | 1687 | |||
| GenBank mRNA | BC089971.1 | - | 2 | 771 | 7315466 | |
More Information
Set Membership: |
|

Gpr19
Genomic Map