siRNA Details
| Gene Symbol: Cops5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025695.1 | NP_001020866.1 | 8 | 1073 | ||
| XM_006237758.2 | XP_006237820.1 | 8 | 1013 | |||
| GenBank mRNA | BC098736.1 | - | 8 | 1073 | 7385252 | |
| FQ214411.1 | - | 8 | 996 | |||
| FQ215612.1 | - | 8 | 996 | |||
| FQ215908.1 | - | 8 | 996 | |||
| FQ227775.1 | - | 8 | 997 | |||
| FQ232542.1 | - | 8 | 996 | |||

Cops5
Genomic Map