siRNA Details
| Gene Symbol: Rb1cc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107901.1 | NP_001101371.1 | 15, 16 | 4275 | ||
| XM_006237794.2 | XP_006237856.1 | 13, 14 | 3865 | |||
| GenBank mRNA | BC091426.1 | - | 1, 2 | 1092 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rb1cc1| Gene Symbol: Rb1cc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107901.1 | NP_001101371.1 | 15, 16 | 4275 | ||
| XM_006237794.2 | XP_006237856.1 | 13, 14 | 3865 | |||
| GenBank mRNA | BC091426.1 | - | 1, 2 | 1092 | ||