siRNA Details
| Gene Symbol: Wdtc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107908.1 | NP_001101378.1 | 7 | 903 | ||
| XM_008764134.1 | XP_008762356.1 | 7 | 1042 | |||
| XM_008764135.1 | XP_008762357.1 | 7 | 699 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdtc1| Gene Symbol: Wdtc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107908.1 | NP_001101378.1 | 7 | 903 | ||
| XM_008764134.1 | XP_008762356.1 | 7 | 1042 | |||
| XM_008764135.1 | XP_008762357.1 | 7 | 699 | |||