siRNA Details
| Gene Symbol: Xkr8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012099.1 | NP_001012099.1 | 3 | 1503 | ||
| GenBank mRNA | AY534262.1 | - | 3 | 1503 | ||
| BC105851.1 | - | 3 | 630 | 7191341 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Xkr8| Gene Symbol: Xkr8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012099.1 | NP_001012099.1 | 3 | 1503 | ||
| GenBank mRNA | AY534262.1 | - | 3 | 1503 | ||
| BC105851.1 | - | 3 | 630 | 7191341 | ||