siRNA Details
| Gene Symbol: Yars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025696.1 | NP_001020867.2 | 4 | 631 | ||
| GenBank mRNA | BC098795.1 | - | 4 | 631 | 7375769 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Yars| Gene Symbol: Yars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025696.1 | NP_001020867.2 | 4 | 631 | ||
| GenBank mRNA | BC098795.1 | - | 4 | 631 | 7375769 | |