siRNA Details
| Gene Symbol: Ythdf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047099.1 | NP_001040564.1 | 5 | 780 | ||
| GenBank mRNA | AB259152.1 | - | 5 | 780 | ||
| BC166407.1 | - | 4 | 695 | 7453874 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ythdf2| Gene Symbol: Ythdf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047099.1 | NP_001040564.1 | 5 | 780 | ||
| GenBank mRNA | AB259152.1 | - | 5 | 780 | ||
| BC166407.1 | - | 4 | 695 | 7453874 | ||