siRNA Details
| Gene Symbol: Tmem67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107916.2 | NP_001101386.3 | 25 | 2601 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem67| Gene Symbol: Tmem67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107916.2 | NP_001101386.3 | 25 | 2601 | ||