siRNA Details
| Gene Symbol: Slc7a13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012100.1 | NP_001012100.1 | 1 | 659 | ||
| GenBank mRNA | BC085796.1 | - | 1 | 659 | 7109760 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc7a13| Gene Symbol: Slc7a13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012100.1 | NP_001012100.1 | 1 | 659 | ||
| GenBank mRNA | BC085796.1 | - | 1 | 659 | 7109760 | |