siRNA Details
| Gene Symbol: Map3k7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107920.2 | NP_001101390.2 | 9 | 1041 | ||
| XM_006237966.2 | XP_006238028.1 | 9 | 2233 | |||
| Gene Symbol: LOC100910771 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750753.3 | XP_003750801.1 | 9 | 1017 | ||
| XM_006245614.2 | XP_006245676.1 | 9 | 1017 | |||

Map3k7
Genomic Map