siRNA Details
| Gene Symbol: Orc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025282.1 | NP_001020453.1 | 7 | 647 | ||
| XM_006237995.2 | XP_006238057.1 | 7 | 655 | |||
| XM_008763621.1 | XP_008761843.1 | 7 | 652 | |||
| XM_008763622.1 | XP_008761844.1 | 7 | 655 | |||
| GenBank mRNA | AB219142.1 | - | 7 | 647 | ||

Orc3
Genomic Map