siRNA Details
Gene Symbol: Zfp462 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006238178.2 | XP_006238240.1 | 5 | 6531 | ||
XM_008763726.1 | XP_008761948.1 | 4 | 11248 | |||
XM_008763727.1 | XP_008761949.1 | 5 | 9236 | |||
XM_008763728.1 | XP_008761950.1 | 4 | 11245 | |||
XM_008763729.1 | XP_008761951.1 | 4 | 11065 |
