siRNA Details
| Gene Symbol: Zfp189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107930.1 | NP_001101400.1 | 3 | 1108 | ||
| XM_006238157.1 | XP_006238219.1 | 4 | 1187 | |||
| XM_008763711.1 | XP_008761933.1 | 3 | 977 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp189| Gene Symbol: Zfp189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107930.1 | NP_001101400.1 | 3 | 1108 | ||
| XM_006238157.1 | XP_006238219.1 | 4 | 1187 | |||
| XM_008763711.1 | XP_008761933.1 | 3 | 977 | |||