siRNA Details
| Gene Symbol: Dfnb31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181088.1 | NP_851602.1 | 2 | 645 | ||
| GenBank mRNA | AY227205.1 | - | 2 | 645 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dfnb31| Gene Symbol: Dfnb31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181088.1 | NP_851602.1 | 2 | 645 | ||
| GenBank mRNA | AY227205.1 | - | 2 | 645 | ||