siRNA Details
| Gene Symbol: Psip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175765.2 | NP_786941.1 | 12 | 1229 | ||
| XM_006238353.2 | XP_006238415.1 | 13 | 1456 | |||
| XM_006238354.2 | XP_006238416.1 | 13 | 1452 | |||
| GenBank mRNA | AB285525.1 | - | 1132 | |||
| AF339084.1 | - | 12 | 1229 | |||

Psip1
Genomic Map