siRNA Details
| Gene Symbol: Klhl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107944.1 | NP_001101414.1 | 1 | 1167 | ||
| GenBank mRNA | BC168717.1 | - | 1 | 1150 | 9102826 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klhl9| Gene Symbol: Klhl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107944.1 | NP_001101414.1 | 1 | 1167 | ||
| GenBank mRNA | BC168717.1 | - | 1 | 1150 | 9102826 | |